Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017242.1 | NP_058938.1 | 1 - 2 | 209 | 123 | |
XM_006248339.2 | XP_006248401.1 | 1 - 2 | 660 | 123 | ||
GenBank mRNA | AY326256.1 | - | 1 - 2 | 178 | 123 | |
BC087607.1 | - | 1 - 2 | 180 | 7314244 | 123 | |
U31554.1 | - | 1 - 2 | 209 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |