Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_218559.8 | XP_218559.4 | 1 - 2 | 1597 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |