Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100559.1 | NP_001094029.1 | 3 - 4 | 382 | 64 | |
GenBank mRNA | BC161911.1 | - | 3 - 4 | 382 | 8368806 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |