Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271072.1 | NP_001258001.1 | 1 - 2 | 284 | 130 | |
GenBank mRNA | FQ212188.1 | - | 1 - 2 | 268 | 130 | |
FQ221944.1 | - | 1 - 2 | 253 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |