Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB195309.1 | - | 6 - 7 | 1334 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |