Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031801.1 | NP_113989.1 | 6 - 7 | 979 | 81 | |
XM_006230582.2 | XP_006230644.1 | 6 - 7 | 1847 | 81 | ||
XM_006230583.2 | XP_006230645.1 | 6 - 7 | 1847 | 81 | ||
XM_008760011.1 | XP_008758233.1 | 6 - 7 | 898 | 81 | ||
XM_008760012.1 | XP_008758234.1 | 6 - 7 | 898 | 81 | ||
XM_008760013.1 | XP_008758235.1 | 6 - 7 | 898 | 81 | ||
XM_008760014.1 | XP_008758236.1 | 6 - 7 | 1863 | 81 | ||
GenBank mRNA | AF055884.1 | - | 6 - 7 | 979 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |