Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglec5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106249.2 | NP_001099719.2 | 7 - 8 | 1462 | 71 | |
XM_006228946.2 | XP_006229008.1 | 7 - 8 | 1578 | 71 | ||
XM_008759327.1 | XP_008757549.1 | 7 - 8 | 1553 | 71 | ||
XM_008759328.1 | XP_008757550.1 | 7 - 8 | 1550 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |