Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107144.1 | NP_001100614.1 | 3 - 4 | 374 | 94 | |
XM_008769232.1 | XP_008767454.1 | 3 - 4 | 335 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |