Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wscd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271178.1 | NP_001258107.1 | 4 - 5 | 943 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |