Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249388.2 | XP_006249450.1 | 1 - 2 | 603 | 67 | |
XM_008769230.1 | XP_008767452.1 | 2 - 3 | 626 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |