Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009683.1 | NP_001009683.1 | 1 - 2 | 253 | 103 | |
GenBank mRNA | BC087678.1 | - | 1 - 2 | 253 | 7318816 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |