Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134358.1 | NP_599185.1 | 6 - 7 | 781 | 95 | |
GenBank mRNA | BC083694.1 | - | 6 - 7 | 1178 | 95 | |
BC158686.1 | - | 6 - 7 | 781 | 8374870 | 95 | |
FQ220404.1 | - | 6 - 7 | 734 | 95 | ||
FQ220639.1 | - | 6 - 7 | 797 | 95 | ||
FQ220740.1 | - | 6 - 7 | 782 | 95 | ||
FQ220961.1 | - | 6 - 7 | 782 | 95 | ||
FQ229512.1 | - | 6 - 7 | 797 | 95 | ||
FQ230125.1 | - | 6 - 7 | 734 | 95 | ||
M29295.1 | - | 5 - 6 | 634 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |