Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013204.1 | NP_001013222.1 | 2 - 3 | 165 | 71 | |
XM_006234859.1 | XP_006234921.1 | 2 - 3 | 260 | 71 | ||
GenBank mRNA | BC091256.1 | - | 2 - 3 | 165 | 7375652 | 71 |
FQ233751.1 | - | 2 - 3 | 201 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |