Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234667.2 | XP_006234729.1 | 4 - 5 | 566 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |