Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107747.2 | NP_001101217.2 | 5 - 6 | 586 | 62 | |
XM_006234551.1 | XP_006234613.1 | 6 - 7 | 623 | 62 | ||
XM_006234552.2 | XP_006234614.1 | 6 - 7 | 653 | 62 | ||
XM_006234553.2 | XP_006234615.1 | 6 - 7 | 623 | 62 | ||
XM_006234554.1 | XP_006234616.1 | 7 - 8 | 665 | 62 | ||
XM_006234556.1 | XP_006234618.1 | 6 - 7 | 600 | 62 | ||
XM_006234557.2 | XP_006234619.1 | 6 - 7 | 630 | 62 | ||
XM_006234558.2 | XP_006234620.1 | 5 - 6 | 571 | 62 | ||
XM_006234559.2 | XP_006234621.1 | 3 - 4 | 2009 | 62 | ||
XM_006234560.2 | XP_006234622.1 | 5 - 6 | 612 | 62 | ||
GenBank mRNA | CK480577.1 | - | 5 - 6 | 586 | 62 | |
EV768858.1 | - | 1 - 2 | 57 | 62 | ||
FQ224002.1 | - | 5 - 6 | 546 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |