Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013119.1 | NP_037251.1 | 8 - 9 | 1446 | 135 | |
XM_008761914.1 | XP_008760136.1 | 7 - 8 | 1249 | 135 | ||
XM_008761915.1 | XP_008760137.1 | 8 - 9 | 1252 | 135 | ||
XM_008761916.1 | XP_008760138.1 | 7 - 8 | 1250 | 135 | ||
XM_008761917.1 | XP_008760139.1 | 7 - 8 | 1249 | 135 | ||
XM_008761918.1 | XP_008760140.1 | 7 - 8 | 1249 | 135 | ||
XM_008761919.1 | XP_008760141.1 | 7 - 8 | 1249 | 135 | ||
XM_008761920.1 | XP_008760142.1 | 7 - 8 | 1249 | 135 | ||
GenBank mRNA | Y00766.1 | - | 8 - 9 | 1446 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |