Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234484.2 | XP_006234546.1 | 7 - 8 | 4779 | 66 | |
XM_006234485.2 | XP_006234547.1 | 7 - 8 | 5259 | 66 | ||
XM_006234486.2 | XP_006234548.1 | 7 - 8 | 4722 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |