Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130422.1 | NP_569106.1 | 8 - 9 | 1183 | 114 | |
XM_008765891.1 | XP_008764113.1 | 9 - 10 | 1264 | 114 | ||
GenBank mRNA | AF317633.1 | - | 8 - 9 | 1183 | 114 | |
FJ465154.1 | - | 9 - 10 | 1246 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |