Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022705.1 | NP_073196.1 | 6 - 7 | 891 | 65 | |
XM_008771886.1 | XP_008770108.1 | 7 - 8 | 1045 | 65 | ||
XM_008771887.1 | XP_008770109.1 | 6 - 7 | 748 | 65 | ||
GenBank mRNA | M16200.1 | - | 6 - 7 | 891 | 65 | |
Y00311.1 | - | 5 - 6 | 626 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |