Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104630.1 | NP_001098100.1 | 4 - 5 | 766 | 75 | |
XM_006254266.2 | XP_006254328.1 | 4 - 5 | 2393 | 75 | ||
GenBank mRNA | AY724491.1 | - | 4 - 5 | 766 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |