Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175757.2 | NP_786933.1 | 6 - 7 | 800 | 81 | |
XM_006252058.2 | XP_006252120.1 | 4 - 5 | 1321 | 81 | ||
GenBank mRNA | AY040223.1 | - | 6 - 7 | 800 | 81 | |
BC078685.1 | - | 6 - 7 | 774 | 7110482 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |