Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106400.1 | NP_001099870.1 | 8 - 9 | 1136 | 129 | |
XM_008760599.1 | XP_008758821.1 | 6 - 7 | 778 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |