Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231845.2 | XP_006231907.1 | 1 - 2 | 268 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |