Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077428.1 | NP_001070896.1 | 4 - 5 | 233 | 161 | |
XM_006231844.2 | XP_006231906.1 | 4 - 5 | 315 | 161 | ||
XM_006231845.2 | XP_006231907.1 | 4 - 5 | 473 | 161 | ||
GenBank mRNA | BC126097.1 | - | 4 - 5 | 233 | 7456682 | 161 |
FQ229200.1 | - | 229 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |