Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100514.1 | NP_001093984.1 | 5 - 6 | 866 | 97 | |
XM_006254858.2 | XP_006254920.1 | 5 - 6 | 1157 | 97 | ||
XM_008772118.1 | XP_008770340.1 | 4 - 5 | 560 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |