Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012189.1 | NP_001012189.1 | 8 - 9 | 917 | 100 | |
GenBank mRNA | BC089869.1 | - | 8 - 9 | 917 | 7324068 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |