Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100647.1 | NP_001094117.1 | 13 - 14 | 1557 | 71 | |
GenBank mRNA | BC160929.1 | - | 13 - 14 | 1557 | 8364923 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |