Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013854.1 | NP_001013876.2 | 2 - 3 | 575 | 70 | |
GenBank mRNA | BC079025.1 | - | 2 - 3 | 575 | 7113945 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |