Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012237.1 | NP_001012237.1 | 3 - 4 | 639 | 135 | |
XM_006247610.2 | XP_006247672.1 | 3 - 4 | 741 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |