Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106720.1 | NP_001100190.1 | 8 - 9 | 888 | 101 | |
XM_006239967.2 | XP_006240029.1 | 12 - 13 | 1169 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |