Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163506.1 | NP_001156978.1 | 5 - 6 | 818 | 109 | |
XM_006239672.2 | XP_006239734.1 | 5 - 6 | 822 | 109 | ||
XM_006239675.2 | XP_006239737.1 | 5 - 6 | 822 | 109 | ||
XM_008764440.1 | XP_008762662.1 | 5 - 6 | 822 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |