Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249138.2 | XP_006249200.1 | 1 - 2 | 174 | 110 | |
GenBank mRNA | M74223.1 | - | 1 - 2 | 170 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |