Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249367.2 | XP_006249429.1 | 2 - 3 | 529 | 84 | |
XM_008769278.1 | XP_008767500.1 | 2 - 3 | 504 | 84 | ||
XM_008769279.1 | XP_008767501.1 | 3 - 4 | 559 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |