Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105938.1 | NP_001099408.1 | 1 | 1238 | 141 | |
XM_006249335.2 | XP_006249397.1 | 2 | 1429 | 141 | ||
XM_006249336.2 | XP_006249398.1 | 2 | 1205 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |