Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mios | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173554.1 | NP_001167025.1 | 10 - 11 | 2707 | 100 | |
XM_006236090.2 | XP_006236152.1 | 9 - 10 | 2707 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |