Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106313.2 | NP_001099783.1 | 3 - 4 | 447 | 128 | |
XM_006230526.2 | XP_006230588.1 | 3 - 4 | 527 | 128 | ||
XM_006230529.2 | XP_006230591.1 | 3 - 4 | 1141 | 128 | ||
XM_008759979.1 | XP_008758201.1 | 3 - 4 | 567 | 128 | ||
XM_008759980.1 | XP_008758202.1 | 3 - 4 | 567 | 128 | ||
XM_008759981.1 | XP_008758203.1 | 3 - 4 | 435 | 128 | ||
GenBank mRNA | BC167006.1 | - | 3 - 4 | 447 | 7314283 | 128 |
EU886468.1 | - | 3 - 4 | 473 | 128 | ||
FQ213357.1 | - | 2 - 3 | 273 | 128 | ||
FQ218084.1 | - | 3 - 4 | 478 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |