Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031779.2 | NP_113967.1 | 9 - 10 | 2316 | 91 | |
XM_006231185.2 | XP_006231247.1 | 7 - 8 | 2416 | 91 | ||
XM_008760312.1 | XP_008758534.1 | 8 - 9 | 2449 | 91 | ||
GenBank mRNA | AF029105.1 | - | 2536 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |