Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col22a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765637.1 | XP_008763859.1 | 60 - 61 | 4852 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |