Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053311.1 | NP_445763.1 | 17 - 18 | 3109 | 97 | |
XM_006241294.2 | XP_006241356.1 | 17 - 18 | 3201 | 97 | ||
XM_006241295.2 | XP_006241357.1 | 16 - 17 | 3078 | 97 | ||
XM_006241296.2 | XP_006241358.1 | 17 - 18 | 3201 | 97 | ||
GenBank mRNA | AF076783.1 | - | 1 - 2 | 70 | 97 | |
J03753.1 | - | 17 - 18 | 3109 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |