Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enthd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242122.2 | XP_006242184.1 | 2 - 3 | 548 | 104 | |
XM_008765787.1 | XP_008764009.1 | 2 - 3 | 548 | 104 | ||
XM_008765788.1 | XP_008764010.1 | 2 - 3 | 548 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |