Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025701.1 | NP_001020872.1 | 8 - 9 | 1143 | 71 | |
GenBank mRNA | BC099200.1 | - | 8 - 9 | 1143 | 7384021 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |