Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242020.2 | XP_006242082.1 | 4 - 5 | 584 | 71 | |
XM_006242021.2 | XP_006242083.1 | 4 - 5 | 584 | 71 | ||
XM_006242022.2 | XP_006242084.1 | 4 - 5 | 735 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |