Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768296.1 | XP_008766518.1 | 7 - 8 | 12668 | 125 | |
GenBank mRNA | BC101875.1 | - | 4 - 5 | 282 | 125 | |
M86514.1 | - | 16041 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |