Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230149.2 | XP_006230211.1 | 3 - 4 | 404 | 85 | |
XM_006230150.1 | XP_006230212.1 | 3 - 4 | 413 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |