Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030585.1 | NP_085074.1 | 6 - 7 | 1003 | 78 | |
GenBank mRNA | BC089795.1 | - | 6 - 7 | 989 | 7375879 | 78 |
U34932.1 | - | 6 - 7 | 1003 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |