Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006971.1 | NP_001006972.1 | 7 - 8 | 613 | 71 | |
XM_006230228.2 | XP_006230290.1 | 8 - 9 | 1310 | 71 | ||
XM_008759871.1 | XP_008758093.1 | 8 - 9 | 1421 | 71 | ||
XM_008759872.1 | XP_008758094.1 | 9 - 10 | 915 | 71 | ||
XM_008759873.1 | XP_008758095.1 | 9 - 10 | 990 | 71 | ||
XM_008759874.1 | XP_008758096.1 | 9 - 10 | 1227 | 71 | ||
XM_008759875.1 | XP_008758097.1 | 9 - 10 | 1367 | 71 | ||
GenBank mRNA | BC083704.1 | - | 7 - 8 | 613 | 7190222 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |