Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023022.2 | NP_075411.2 | 8 - 9 | 1238 | 67 | |
GenBank mRNA | AB015194.1 | - | 8 - 9 | 1211 | 67 | |
AF531097.1 | - | 8 - 9 | 1238 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |