Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033707.1 | NP_001028879.1 | 7 - 8 | 657 | 82 | |
GenBank mRNA | BC101917.1 | - | 7 - 8 | 657 | 7935829 | 82 |
FQ218185.1 | - | 7 - 8 | 729 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |