Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234484.2 | XP_006234546.1 | 2 - 3 | 258 | 119 | |
XM_006234485.2 | XP_006234547.1 | 2 - 3 | 738 | 119 | ||
XM_006234486.2 | XP_006234548.1 | 2 - 3 | 201 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |