Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005903.1 | NP_001005903.1 | 3 - 4 | 297 | 93 | |
XM_006239171.2 | XP_006239233.1 | 4 - 5 | 1349 | 93 | ||
XM_006239172.2 | XP_006239234.1 | 4 - 5 | 1346 | 93 | ||
XM_006239174.2 | XP_006239236.1 | 3 - 4 | 1276 | 93 | ||
XM_006239175.2 | XP_006239237.1 | 3 - 4 | 1272 | 93 | ||
XM_006239177.2 | XP_006239239.1 | 3 - 4 | 326 | 93 | ||
GenBank mRNA | BC083861.1 | - | 3 - 4 | 297 | 7126848 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |